Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIAA0284 Mouse anti-Human, Clone: 1D4B8, Proteintech
SDP

Catalog No. 501737157 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-7157 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-7157 Supplier Proteintech Group Inc Supplier No. 667491IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CEP170B plays a role in microtubule organization.

Specifications

Antigen KIAA0284
Applications Western Blot
Classification Monoclonal
Clone 1D4B8
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene CEP170B
Gene Accession No. Q9Y4F5
Gene Alias Cep170B, FAM68C, KIAA0284, Protein KIAA0284
Gene Symbols CEP170B
Host Species Mouse
Immunogen KIAA0284 Fusion Protein Ag26852
Purification Method Protein A
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 283638
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.