Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIAA0319 Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NB10093472
Description
KIAA0319 Polyclonal specifically detects KIAA0319 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
KIAA0319 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
DLX2, DYLX2, dyslexia susceptibility 2, DYX2, KIAA0319, MGC176717 | |
Goat | |
Affinity Purified | |
RUO | |
9856 | |
Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Western Blot 0.5 - 1.5 ug/ml, Immunohistochemistry 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500, Peptide ELISA Detection limit 1:128000 | |
NP_001074520.1 | |
KIAA0319 | |
Peptide with sequence C-QGKIKQENKPTLH corresponding to internal region according to Uniprot Mouse NP_001074520.1. | |
0.1 mg | |
Primary | |
The variants XP_341528.2 and XP_001070349.1 represent identical protein. This antibody is expected to cross-react with mouse KIAA0319 isoform 1 (NP_001074520.1), isoform 2 (XP_994023.1) and isoform 4 (XP_913491.1). | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction