Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kiaa0319 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553339
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553339 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553339 Supplier Invitrogen™ Supplier No. PA553339
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GRTYSNAVIS PNLETTRIMR VSHTFPVVDC TAACCDLSSC DLAWWFEGRC YLVSCPHKEN CEPKKMGPIR SYLTFVLRPV QRPAQLLDYG DMMLNRGSPS GIWGDSPEDI RKDLPFLGKD WGLEEMSEYS DDYRELEKDL LQPSG Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 79%.

This gene encodes a transmembrane protein that contains a large extracellular domain with multiple polycystic kidney disease (PKD) domains. The encoded protein may play a role in the development of the cerebral cortex by regulating neuronal migration and cell adhesion. Single nucleotide polymorphisms in this gene are associated with dyslexia. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. [provided by RefSeq, Nov 2011].
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kiaa0319
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KIAA0319
Gene Accession No. Q5VV43
Gene Alias 4930451E12Rik; D130043K22Rik; DYLX2; dyslexia susceptibility 2; dyslexia-associated protein KIAA0319; Dyslexia-associated protein KIAA0319 homolog; DYX2; Kiaa0319; LOW QUALITY PROTEIN: dyslexia-associated protein KIAA0319 homolog; mKIAA0319; neuronal migration; NMIG; RIKEN cDNA D130043K22 gene
Gene Symbols KIAA0319
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human Kiaa0319. Recombinant protein control fragment (Product #RP-90263).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9856
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.