Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA0319 Polyclonal Antibody

Catalog No. PIPA590416
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590416 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590416 Supplier Invitrogen™ Supplier No. PA590416

Rabbit Polyclonal Antibody

Immunogen sequence: ARKQCSEGRT YSNAVISPNL ETTRIMRVSH TFPVVDCTAA CCDLSSCDLA WWFEGRCYLV SCPHKENCEP KKMGPIRSYL TFVLRPVQRP AQLLDYGDMM LNRGSPSGIW GDSPEDIRKD LTFLGKDWGL EEMSEYSDDY RELEKDLLQP SGKQEPRGSA EYTDWGLLPG SEGAFNSSVG DSPAVPAETQ QDPELHYLNE SASTPAPKLP ERSVLLPLPT TPSSGEVLEK EKASQLQEQS SNSSGKEVLM PSHSLPPASL ELSSVTVEKS PVLTVTPGST; Positive Samples: U-87MG, Mouse brain; Cellular Location: Cell membrane, Early endosome membrane, Single-pass type I membrane protein.

This gene encodes a transmembrane protein that contains a large extracellular domain with multiple polycystic kidney disease (PKD) domains. The encoded protein may play a role in the development of the cerebral cortex by regulating neuronal migration and cell adhesion. Single nucleotide polymorphisms in this gene are associated with dyslexia. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. [provided by RefSeq, Nov 2011].
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA0319
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene KIAA0319
Gene Accession No. Q5SZV5, Q5VV43
Gene Alias 4930451E12Rik; D130043K22Rik; DYLX2; dyslexia susceptibility 2; dyslexia-associated protein KIAA0319; Dyslexia-associated protein KIAA0319 homolog; DYX2; Kiaa0319; LOW QUALITY PROTEIN: dyslexia-associated protein KIAA0319 homolog; mKIAA0319; neuronal migration; NMIG; RIKEN cDNA D130043K22 gene
Gene Symbols D130043K22Rik, KIAA0319
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 21-300 of human KIAA0319 (NP_0556242).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 210108, 9856
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.