Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIAA0319 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30485420UL
Description
KIAA0319 Polyclonal specifically detects KIAA0319 in Human, Mouse samples. It is validated for Western Blot.Specifications
KIAA0319 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
9856 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
DLX2, DYLX2, dyslexia susceptibility 2, DYX2, KIAA0319, MGC176717 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 21-300 of human KIAA0319 (NP_055624.2). | |
20 μL | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction