Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIAA0319L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567257
Description
Immunogen sequence: KRKSKYKILDA TDQESLELKP TSRAGIKQKG LLLSSSLMHS ESELDSDDAI FTWPDREKGK LLHGQNGSVP NGQ Highest antigen sequence identity to the following orthologs - mouse 93%, rat 93%.
Possible role in axon guidance through interaction with RTN4R. (Microbial infection) Acts as a receptor for adeno-associated virus and is involved in adeno-associated virus infection through endocytosis system.
Specifications
KIAA0319L | |
Polyclonal | |
Unconjugated | |
KIAA0319L | |
A730047D20Rik; AAV receptor; AAVR; Adeno-associated virus receptor; AU040320; Dyslexia-associated protein KIAA0319-like protein; dyslexia-associated protein KIAA0319-like protein homolog; expressed sequence AU040320; KIAA0319 like; Kiaa0319l; KIAA0319-like; KIAA0319-like ortholog; KIAA1837; likely ortholog of H. sapiens polycystic kidney disease 1-like (PKD1-like); LOW QUALITY PROTEIN: dyslexia-associated protein KIAA0319-like protein homolog; polycystic kidney disease 1-related; PP791 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79932 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8IZA0 | |
KIAA0319L | |
Recombinant Human KIAA0319L. Recombinant protein control fragment (Product #RP-107893). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction