Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA0319L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA569645
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA569645 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA569645 Supplier Invitrogen™ Supplier No. PA569645
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.

Possible role in axon guidance through interaction with RTN4R. (Microbial infection) Acts as a receptor for adeno-associated virus and is involved in adeno-associated virus infection through endocytosis system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA0319L
Applications Western Blot, Surface Plasmon Resonance
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene KIAA0319L
Gene Accession No. Q8IZA0
Gene Alias A730047D20Rik; AAV receptor; AAVR; Adeno-associated virus receptor; AU040320; Dyslexia-associated protein KIAA0319-like protein; dyslexia-associated protein KIAA0319-like protein homolog; expressed sequence AU040320; KIAA0319 like; Kiaa0319l; KIAA0319-like; KIAA0319-like ortholog; KIAA1837; likely ortholog of H. sapiens polycystic kidney disease 1-like (PKD1-like); LOW QUALITY PROTEIN: dyslexia-associated protein KIAA0319-like protein homolog; polycystic kidney disease 1-related; PP791
Gene Symbols KIAA0319L
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human KIAA0319L.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79932
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.