Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIAA0323 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551923
Description
Immunogen sequence: PFTFVGNLFM VPDDPLGRNG PTLDEFLKKP ARTQGSSKAQ HPSRGFAEHG KQQQGREEEK GSGGIRKTRE TERLRRQLLE VFWGQDHKVD FILQREPYCR DINQ Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 90%.
The KIAA0323 gene, also known as chromosome 18 open reading frame 8, encodes a protein of currently undefined function. Located on chromosome 18 at the 18q21.1 position, KIAA0323 is part of a family of genes that were initially identified through the sequencing of human brain cDNA libraries. Its precise role in cellular processes remains unclear, but like other KIAA proteins, it may be involved in various cellular functions such as signaling pathways, structural organization, and regulatory mechanisms. Further research is required to elucidate the exact biological pathways in which KIAA0323 participates and its potential implications in human health and disease.
Specifications
KIAA0323 | |
Polyclonal | |
Unconjugated | |
KHNYN | |
6330555F21; 9130227C08Rik; AI605259; KH and NYN domain containing; KH and NYN domain-containing protein; KHNYN; KIAA0323; mKIAA0323; Protein KHNYN | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23351 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O15037 | |
KHNYN | |
Recombinant protein corresponding to Human KIAA0323. Recombinant protein control fragment (Product #RP-89122). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction