Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA0323 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531660
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531660 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531660 Supplier Invitrogen™ Supplier No. PA531660
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HCT116, mouse brain. Predicted reactivity: Mouse (86%), Bovine (90%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The KIAA0323 gene, also known as chromosome 18 open reading frame 8, encodes a protein of currently undefined function. Located on chromosome 18 at the 18q21.1 position, KIAA0323 is part of a family of genes that were initially identified through the sequencing of human brain cDNA libraries. Its precise role in cellular processes remains unclear, but like other KIAA proteins, it may be involved in various cellular functions such as signaling pathways, structural organization, and regulatory mechanisms. Further research is required to elucidate the exact biological pathways in which KIAA0323 participates and its potential implications in human health and disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA0323
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.59 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene KHNYN
Gene Accession No. O15037, Q80U38
Gene Alias 6330555F21; 9130227C08Rik; AI605259; KH and NYN domain containing; KH and NYN domain-containing protein; KHNYN; KIAA0323; mKIAA0323; Protein KHNYN
Gene Symbols KHNYN
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human KIAA0323. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219094, 23351
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.