Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA0391 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100619
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100619 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100619 Supplier Invitrogen™ Supplier No. PA5100619
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MRPP3.

KIAA0391, also known as MRPP3 (mitochondrial ribonuclease P protein 3), is a 583 amino acid protein that is located in the mitochondria. KIAA0391 functions in tRNA maturation and is a component of mitochondrial ribonuclease P which cleaves tRNA at the 5'-end. Mitochondrial ribonuclease P is an enzyme composed of RG9MTD1, MRPP22 and KIAA0391 subunits. Due to alternative splicing events, KIAA0391 exists as four isoforms. The gene encoding KIAA0391 maps to human chromosome 14, which contains about 700 genes and makes up about 3. 5% of human cellular DNA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA0391
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PRORP
Gene Accession No. B5DF07, O15091, Q8JZY4
Gene Alias 1110008L16Rik; KIAA0391; Mitochondrial ribonuclease P catalytic subunit; Mitochondrial ribonuclease P protein 3; mitochondrial RNase P protein 3; mitochondrial RNase P subunit 3; mKIAA0391; MRPP3; Prorp; Protein only RNase P catalytic subunit; proteinaceous RNase P; RGD1305089; RIKEN cDNA 1110008L16 gene; similar to 1110008L16Rik protein
Gene Symbols PRORP
Host Species Rabbit
Immunogen A synthesized peptide derived from human KIAA0391(Accession O15091), corresponding to amino acid residues I292-V342.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 299050, 66132, 9692
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.