Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA0430 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553696
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553696 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553696 Supplier Invitrogen™ Supplier No. PA553696
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PPVCLKGSLY CEDCLNKPAR NSIIDAAKVW PNIPPPNTQP APLAVPLCNG CGTKGTGKET TLLLATSLGK AASKFGSPEV AVAGQVLENL PPIGVFWDIE NCSVPSGRSA TAVVQRIREK FFKGHREAEF ICVCDISKEN KE Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.

This gene encodes a putative peroxisomal protein that appears to be conserved across Euteleostomi. In humans, it may be autoantigenic.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA0430
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MARF1
Gene Accession No. Q9Y4F3
Gene Alias 4921513D23Rik; BC031575; C87306; E030019O05; KIAA0430; limkain b1; limkain-b1; Lkap; MARF1; meiosis arrest female 1; meiosis arrest female protein 1; Meiosis regulator and mRNA stability factor 1; mKIAA0430; PPP1R34; protein phosphatase 1, regulatory subunit 34
Gene Symbols MARF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KIAA0430. Recombinant protein control fragment (Product #RP-91023).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9665
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.