Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIAA0513 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546026
Description
Peptide sequence: NVKGFFGGLE TKLKGPLARR NEEDENKPQE KRPRAVTAYS PEDEKKGEKI Sequence homology: Horse: 82%; Human: 100%.
The function of this protein remains unknown.
Specifications
| KIAA0513 | |
| Polyclonal | |
| Unconjugated | |
| KIAA0513 | |
| 6430548M08Rik; AW049007; KIAA0513; mKIAA0513; RGD1304884; RIKEN cDNA 6430548M08 gene; similar to RIKEN cDNA 6430548M08; uncharacterized protein KIAA0513; uncharacterized protein KIAA0513 homolog | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 9764 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| O60268 | |
| KIAA0513 | |
| Synthetic peptide directed towards the C-terminal of human KIAA0513 (aa 240-289). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction