Learn More
Invitrogen™ KIAA0513 Polyclonal Antibody

Description
KIAA0513 Polyclonal Antibody for Western Blot, ICC/IF, ELISA
Specifications
Specifications
| Antigen | KIAA0513 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 4 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | KIAA0513 |
| Gene Accession No. | O60268, Q8R0A7 |
| Gene Alias | 6430548M08Rik; AW049007; KIAA0513; mKIAA0513; RGD1304884; RIKEN cDNA 6430548M08 gene; similar to RIKEN cDNA 6430548M08; uncharacterized protein KIAA0513; uncharacterized protein KIAA0513 homolog |
| Gene Symbols | 6430548M08Rik, KIAA0513 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.