Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIAA0513 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501989829
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-9829 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-9829 Supplier Bioss Supplier No. BS16975R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The function of this protein remains unknown.

Specifications

Antigen KIAA0513
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation TBS with 1% BSA, 50% glycerol and 0.03% Proclin 300; pH
Gene KIAA0513
Gene Accession No. O60268
Gene Alias 6430548M08Rik; AW049007; Kiaa0513; mKIAA0513; RGD1304884; RIKEN cDNA 6430548M08 gene; similar to RIKEN cDNA 6430548M08; Uncharacterized protein KIAA0513; uncharacterized protein KIAA0513 homolog
Gene Symbols KIAA0513
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human KIAA0513.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9764
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.