Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA0701 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558875
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558875 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558875 Supplier Invitrogen™ Supplier No. PA558875
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SVNAGANLQN YGETSPDAIS TNSEGAQENH DDLMSVVVFK ITGVNGEIDI RGEDTEICLQ VNQVTPDQLG NISLRHYLCN RPVGSDQKAV IHS Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 78%.

UHRF1BP1L gene ontology annotations related to this gene include GARP complex binding; protein homodimerization activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA0701
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Uhrf1bp1l
Gene Accession No. A0JNW5
Gene Alias 2010319N22Rik; 4930506D01Rik; AA589495; C80672; E030041M21Rik; Kiaa0701; mKIAA0701; SHIP164; syntaxin 6Habc-interacting protein of 164 kDa; Syntaxin-6 Habc-interacting protein of 164 kDa; UHRF1 (ICBP90) binding protein 1-like; UHRF1 binding protein 1 like; UHRF1 binding protein 1-like; UHRF1-binding protein 1-like; UHRF1BP1L
Gene Symbols Uhrf1bp1l
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KIAA0701. Recombinant protein control fragment (Product #RP-97708).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23074
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.