Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIAA0895 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA554313
Description
Immunogen sequence: AIKPTNLEKE KLRFFKSDYT YNPQFEYANP ALPSVLAKHS HASDRFLKQI VVHLTEDLLS RASMTVVNGC PTLTINVSTA R Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 60%.
KIAA0895 is a protein coding gene.
Specifications
KIAA0895 | |
Polyclonal | |
Unconjugated | |
KIAA0895 | |
1110003N12Rik; 9530077C05Rik; KIAA0895; mKIAA0895; RIKEN cDNA 9530077C05 gene; uncharacterized protein KIAA0895 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23366 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NCT3 | |
KIAA0895 | |
Recombinant protein corresponding to Human KIAA0895. Recombinant protein control fragment (Product #RP-92788). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction