Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA0895 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584477
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584477 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584477 Supplier Invitrogen™ Supplier No. PA584477
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KLHWPEQELA KKSILNAEDS LIIDNKRSIS HLSSGVLKDI FTTGTSSYNV LLQSKEEKKY HSQKQSSSTY SKRCRKPSKS PNTSRSKDP Highest antigen sequence indentity to the following orthologs: Mouse - 75%, Rat - 75%.

KIAA0895 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA0895
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KIAA0895
Gene Accession No. Q8NCT3
Gene Alias 1110003N12Rik; 9530077C05Rik; KIAA0895; mKIAA0895; RIKEN cDNA 9530077C05 gene; uncharacterized protein KIAA0895
Gene Symbols KIAA0895
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KIAA0895. Recombinant protein control fragment (Product #RP-107041).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23366
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.