Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIAA0907/BLOM7 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501569121
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-9121 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-9121 Supplier Bethyl Laboratories, Inc Supplier No. A304956A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse

RNA-binding protein involved in pre-mRNA splicing. Interacts with the PRP19C/Prp19 complex/NTC/Nineteen complex which is part of the spliceosome. Involved in regulating splice site selection. Binds preferentially RNA with A/C rich sequences and poly-C stretches.

Specifications

Antigen KIAA0907/BLOM7
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene KHDC4
Gene Accession No. Q7Z7F0
Gene Alias BLOM7
Gene Symbols KHDC4
Host Species Rabbit
Immunogen Between 564 and 614
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22889
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.