Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIAA0907 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552612
Description
Immunogen sequence: KGKLKPTQNA SEKLQAPGKG LTSNKSKDDL VVAEVEINDV PLTCRNLLTR GQTQDEISRL SGAAVSTRGR FMTTEEKAKV GPGDRPLYLH VQGQTRELVD RAVNRIKEII TNGVVKAATG TSPTFNGATV TVYHQPAP Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
RNA-binding protein involved in pre-mRNA splicing (PubMed:19641227). Interacts with the PRP19C/Prp19 complex/NTC/Nineteen complex which is part of the spliceosome (PubMed:19641227). Involved in regulating splice site selection (PubMed:19641227). Binds preferentially RNA with A/C rich sequences and poly-C stretches (PubMed:23144703). [UniProt].
Specifications
| KIAA0907 | |
| Polyclonal | |
| Unconjugated | |
| KHDC4 | |
| 2810403A07Rik; A430106P18Rik; AI256352; AI451678; BLOM7; Brings lots of money 7; KH homology domain-containing protein 4; Khdc4; KIAA0907; Pre-mRNA splicing factor protein KHDC4; RIKEN cDNA 2810403A07 gene; SNORA80EHG; UPF0469 protein KIAA0907 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22889 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z7F0 | |
| KHDC4 | |
| Recombinant protein corresponding to Human KIAA0907. Recombinant protein control fragment (Product #RP-89154). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction