Learn More
KIAA0947 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304276AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Component of the little elongation complex (LEC), a complex required to regulate small nuclear RNA (snRNA) gene transcription by RNA polymerase II and III (PubMed:22195968, PubMed:23932780). Specifically acts as a scaffold protein that promotes the LEC complex formation and recruitment and RNA polymerase II occupancy at snRNA genes in subnuclear bodies (PubMed:23932780).Specifications
KIAA0947 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9Y2F5 | |
ICE1 | |
Between 1582 and 1632 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ICE1 | |
interactor of little elongator complex ELL subunit 1, KIAA0947 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23379 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.