Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIAA1024 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552839
Description
Immunogen sequence: IQSSCYNSTG SLSQLHKSDC DSSPEHNLTK IANGVPNSKG DKGNRPENTH HSEEELKTSV CKLVLRIGEI ERKLESLSGV RDEISQVLGK LNKLDQKMQQ PEKVSVQIDL NSLTSEGPSD DSASPRMFHA HS Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 90%.

Specifications
KIAA1024 | |
Polyclonal | |
Unconjugated | |
MINAR1 | |
AF529169; cDNA sequence AF529169; DD1; KIAA1024; Major intrinsically disordered Notch2-binding receptor 1; Membrane integral NOTCH2-associated receptor 1; MINAR1; Protein DD1; Ubiquitination and mTOR signaling protein; Ubtor; UPF0258 protein KIAA1024 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23251 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UPX6 | |
MINAR1 | |
Recombinant protein corresponding to Human KIAA1024. Recombinant protein control fragment (Product #RP-90443). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction