Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIAA1033 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561064
Description
Immunogen sequence: KVLNKVITVY AALCCEIKKL KYEAETKFYN GLLFYGEGAT DASMVEGDCQ IQMGRFISFL QELSCFVTRC YEVVMNVVHQ LAALYISNKI APKII Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 91%.
This gene encodes a component of the WASH complex, which functions in the intracellular transport of endosomes. Mutations in this gene have been detected in individuals with autosomal recessive cognitive disability. Alternative splicing results in multiple transcript variants.
Specifications
KIAA1033 | |
Polyclonal | |
Unconjugated | |
WASHC4 | |
A230046K03Rik; AA589518; Kiaa1033; mKIAA1033; MRT43; RIKEN cDNA A230046K03 gene; strumpellin and WASH-interacting protein; SWIP; WASH complex subunit 4; WASH complex subunit 7; WASH complex subunit SWIP; WASHC4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23325 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q2M389 | |
WASHC4 | |
Recombinant protein corresponding to Human KIAA1033. Recombinant protein control fragment (Product #RP-97753). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction