Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA1045 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554290
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554290 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554290 Supplier Invitrogen™ Supplier No. PA554290
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLTEEEMYSL TETFQRCKVI PDCSLTLEDF LRYRHQAAKR GDRDRALSEE QEEQAARQFA ALDPEHRGHI EWPDFLSHES LLLLQQLR Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 94%.

PHF24 gene ontology annotations related to this gene include detection of mechanical stimulus involved in sensory perception of pain; gamma-aminobutyric acid signaling pathway; regulation of G protein-coupled receptor signaling pathway; regulation of synaptic transmission, GABAergic.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA1045
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PHF24
Gene Accession No. Q9UPV7
Gene Alias A230025O18; expressed sequence N28178; Kiaa1045; mKIAA1045; N28178; PHD finger protein 24; Phf24; protein KIAA1045; RGD1559864; RP11-392A14.4; similar to mKIAA1045 protein
Gene Symbols PHF24
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KIAA1045. Recombinant protein control fragment (Product #RP-92938).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23349
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.