Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA1045 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571131
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571131 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571131 Supplier Invitrogen™ Supplier No. PA571131
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%.

PHF24 gene ontology annotations related to this gene include detection of mechanical stimulus involved in sensory perception of pain; gamma-aminobutyric acid signaling pathway; regulation of G protein-coupled receptor signaling pathway; regulation of synaptic transmission, GABAergic.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA1045
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene PHF24
Gene Accession No. Q9UPV7
Gene Alias A230025O18; expressed sequence N28178; Kiaa1045; mKIAA1045; N28178; PHD finger protein 24; Phf24; protein KIAA1045; RGD1559864; RP11-392A14.4; similar to mKIAA1045 protein
Gene Symbols PHF24
Host Species Rabbit
Immunogen synthetic peptide directed towards the N-terminal region of human KIAA1045.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23349
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.