Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA1143 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566424
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566424 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566424 Supplier Invitrogen™ Supplier No. PA566424
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MSKRNQVSYVR PAEPAFLARF KERVGYREGP TVETKRIQPQ P Highest antigen sequence identity to the following orthologs - mouse 90%, rat 93%.

KIAA1143 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA1143
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KIAA1143
Gene Accession No. Q96AT1
Gene Alias KIAA1143; Uncharacterized protein KIAA1143
Gene Symbols KIAA1143
Host Species Rabbit
Immunogen Recombinant Human KIAA1143. Recombinant protein control fragment (Product #RP-105210).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57456
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.