Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIAA1161 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566926
Description
Immunogen sequence: DSVPFHLGWNS TERSLRLQAR YHDTPYKPPA GRAAAPELSY RVCVGSDVTS IHKYMVRRYF NKPSRVPAPE AFRDPIW Highest antigen sequence identity to the following orthologs - mouse 95%, rat 94%.
Putative glycosidase. Promotes myogenesis by activating AKT signaling through the maturation and secretion of IGF2. [UniProt].
Specifications
KIAA1161 | |
Polyclonal | |
Unconjugated | |
MYORG | |
KIAA1161; Myogenesis-regulating glycosidase; MYORG; NET37; uncharacterized family 31 glucosidase KIAA1161 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57462 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6NSJ0 | |
MYORG | |
Recombinant Human KIAA1161. Recombinant protein control fragment (Product #RP-107578). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction