Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA1191 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560798
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560798 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560798 Supplier Invitrogen™ Supplier No. PA560798
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ALEASAPLGK MSLPIGIYRR AVSYDDTLED PAPMTPPPSD MGSVPWKPVI PERKYQHLAK VEEGEASLPS PAMTLSSAID SVDKVPVVKA KATHVIMNSL IT Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 78%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA1191
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KIAA1191
Gene Accession No. Q96A73
Gene Alias 4833439L19Rik; 4930558H15Rik; brain-derived rescue factor p60MONOX; C81457; flavin monooxygenase motif-containing protein of 33 kDa; flavine monooxygenase motif-containing protein of 33 kDa; KIAA1191; P33monox; p60MONOX; Putative monooxygenase p33MONOX; RIKEN cDNA 4833439L19 gene
Gene Symbols KIAA1191
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KIAA1191. Recombinant protein control fragment (Product #RP-100122).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57179
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.