Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA1191 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113234
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113234 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113234 Supplier Invitrogen™ Supplier No. PA5113234
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KIAA1191 Polyclonal Antibody for Western Blot, IHC (P), ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA1191
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene KIAA1191
Gene Accession No. Q96A73, Q9DBN4
Gene Alias 4833439L19Rik; 4930558H15Rik; brain-derived rescue factor p60MONOX; C81457; flavin monooxygenase motif-containing protein of 33 kDa; flavine monooxygenase motif-containing protein of 33 kDa; KIAA1191; P33monox; p60MONOX; Putative monooxygenase p33MONOX; RIKEN cDNA 4833439L19 gene
Gene Symbols 4833439L19Rik, KIAA1191
Host Species Rabbit
Immunogen Recombinant Human Putative monooxygenase p33MONOX protein (32-305AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57179, 97820
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.