Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA1199 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA524444
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA524444 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA524444 Supplier Invitrogen™ Supplier No. PA524444
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

KIAA1199 Polyclonal Antibody for Western Blot

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA1199
Applications Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Cemip
Gene Accession No. Q8BI06, Q8WUJ3
Gene Alias 12H19.01.T7; 6330404C01Rik; 9930013L23Rik; AY007814; CCSP1; cell migration inducing hyaluronan binding protein; cell migration inducing hyaluronidase 1; cell migration inducing protein, hyaluronan binding; cell migration-inducing and hyaluronan-binding protein; CEMIP; colon cancer secreted protein 1; gb:bi851057; hyaluronan-binding protein involved in hyaluronan depolymerization; HYBID; Kiaa1199; Protein 12H19.01.T7; protein KIAA1199; RGD1305254; TMEM2L
Gene Symbols Cemip
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 1292-1321 amino acids from the C-terminal region of human KIAA1199.
Purification Method Antigen Affinity Chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57214, 80982
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.