Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIAA1199 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA5106288
Description
Antibody detects endogenous levels of total KIAA1199.
The function of this protein remains unknown.
Specifications
KIAA1199 | |
Polyclonal | |
Unconjugated | |
Cemip | |
12H19.01.T7; 6330404C01Rik; 9930013L23Rik; AY007814; CCSP1; cell migration inducing hyaluronan binding protein; cell migration inducing hyaluronidase 1; cell migration inducing protein, hyaluronan binding; cell migration-inducing and hyaluronan-binding protein; CEMIP; colon cancer secreted protein 1; gb:bi851057; hyaluronan-binding protein involved in hyaluronan depolymerization; HYBID; Kiaa1199; Protein 12H19.01.T7; protein KIAA1199; RGD1305254; TMEM2L | |
Rabbit | |
Affinity chromatography | |
RUO | |
308797, 57214, 80982 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8BI06, Q8WUJ3 | |
Cemip | |
A synthesized peptide derived from human CEMIP(Accession Q8WUJ3), corresponding to amino acid residues K1106-M1156. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction