Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIAA1211 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA560292
Description
Immunogen sequence: EEKREEGDTE PLLKQEGPVE AAQPPVERKE AAALEQGRKV EELRWQEVDE RQTMPRPYTF QVSSGGKQIL FPKVNLSPVT PAKDTGLTAA PQEPKAPKAS PVQ Highest antigen sequence identity to the following orthologs: Mouse - 61%, Rat - 53%.

Specifications
KIAA1211 | |
Polyclonal | |
Unconjugated | |
KIAA1211 | |
Cancer-related regulator of actin dynamics; CRAD; KIAA1211; uncharacterized protein KIAA1211 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57482 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6ZU35 | |
KIAA1211 | |
Recombinant protein corresponding to Human KIAA1211. Recombinant protein control fragment (Product #RP-100110). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction