Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIAA1211L Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566411
Description
Immunogen sequence: LREAAEGLGED STGKKKSKFK TFKKFFGKKK RKESPSSTGS STWKQSQTRN EVIAIESGPV GYDSEDELEE SR Highest antigen sequence identity to the following orthologs - mouse 81%, rat 80%.
KIAA1211L is a protein coding gene.
Specifications
| KIAA1211L | |
| Polyclonal | |
| Unconjugated | |
| KIAA1211L | |
| C2orf55; KIAA1211 like; KIAA1211L; KIAA1211-like; uncharacterized protein C2orf55; uncharacterized protein KIAA1211-like | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 343990 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6NV74 | |
| KIAA1211L | |
| Recombinant Human KIAA1211L. Recombinant protein control fragment (Product #RP-105200). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction