Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA1217 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585468
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585468 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585468 Supplier Invitrogen™ Supplier No. PA585468
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

KIAA1217 required for normal development of intervertebral disks.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA1217
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.89 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene KIAA1217
Gene Accession No. Q5T5P2
Gene Alias KIAA1217; likely orthologue of Mus musculus enhancer trap locus 4; RP11-324E23.1; sickle tail protein homolog; SKT
Gene Symbols KIAA1217
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human SKT. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56243
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.