Learn More
KIAA1217/SKT Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304928AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Required for normal development of intervertebral disks.Specifications
KIAA1217/SKT | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q5T5P2 | |
KIAA1217 | |
Between 1625 and 1675 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
KIAA1217 | |
RP11-324E23.1, SKT | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56243 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.