Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA1310 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565136
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565136 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565136 Supplier Invitrogen™ Supplier No. PA565136
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ISSPVYSESDV PIDVETVTST PMPLYDNQKA RSVMNECERH VIFARTDADA PPPPEDWEEH VNRTGWTMAQ NKLFNKIL Highest antigen sequence identity to the following orthologs - mouse 87%, rat 87%.

As part of the NSL complex it is involved in acetylation of nucleosomal histone H4 on several lysine residues and therefore may be involved in the regulation of transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA1310
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KANSL3
Gene Accession No. Q9P2N6
Gene Alias KANSL3; KAT8 regulatory NSL complex subunit 3; KIAA1310; Non-specific lethal 3 homolog; NSL complex protein NSL3; NSL3; PRTD; Rcd1; serum inhibited-related protein; SI1; Testis development protein PRTD
Gene Symbols KANSL3
Host Species Rabbit
Immunogen Recombinant Human KIAA1310. Recombinant protein control fragment (Product #RP-105895).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55683
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.