Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA1310 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596979
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596979 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596979 Supplier Invitrogen™ Supplier No. PA596979
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MAHRGGERDF QTSARRMGTS LLFQLSVHER ELDLVFLDHS YAKPWSAHPD ASSARPTRML FVTPRRQHES TIESDVPIDV ETVTSTPMPL YDNQKARSVM NECERHVIFA RTDADAPPPP EDWEEHVNRT GWTMAQNKLF NKILKALQSD RLARLANEGA CNEPVLRRVA VDKCARRVRQ ALASVSWDTK LIQWLHTTLV; Positive Samples: U-251MG, Jurkat; Cellular Location: Nucleus.

As part of the NSL complex it is involved in acetylation of nucleosomal histone H4 on several lysine residues and therefore may be involved in the regulation of transcription.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA1310
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene KANSL3
Gene Accession No. Q9P2N6
Gene Alias KANSL3; KAT8 regulatory NSL complex subunit 3; KIAA1310; Non-specific lethal 3 homolog; NSL complex protein NSL3; NSL3; PRTD; Rcd1; serum inhibited-related protein; SI1; Testis development protein PRTD
Gene Symbols KANSL3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human KANSL3 (NP_0011084881).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55683
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.