Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA1324 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567123
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567123 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567123 Supplier Invitrogen™ Supplier No. PA567123
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELPHGFASLSA NMELDDSAAE STGNCTSSKW VPRGDYIASN TDECTATLMY AVNLKQSGTV NFEYYYPDSS I Highest antigen sequence identity to the following orthologs - mouse 93%, rat 93%.

Expression of this gene is induced by estrogen and the encoded protein has been characterized as a transmembrane protein. The encoded protein has been found in to correlate with survival in certain carcinomas (PMID: 21102415) and may be important for cellular response to stress (PMID: 21072319). Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA1324
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KIAA1324
Gene Accession No. Q6UXG2
Gene Alias EIG121; Estrogen-induced gene 121 protein; KIAA1324; maba1; UNQ2426/PRO4985; UPF0577 protein KIAA1324
Gene Symbols KIAA1324
Host Species Rabbit
Immunogen Recombinant Human KIAA1324. Recombinant protein control fragment (Product #RP-107769).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57535
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.