Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA1324L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567405
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567405 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567405 Supplier Invitrogen™ Supplier No. PA567405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YFHFFNISLCG HEGKKMALCT NNITDFTVKE IVAGSDDYTN LVGAFVCQST IIPSESKGFR AALSSQSIIL ADTFIGVTVE TT Highest antigen sequence identity to the following orthologs - mouse 90%, rat 89%.

KIAA1324L is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA1324L
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KIAA1324L
Gene Accession No. A8MWY0
Gene Alias 9330182L06Rik; EIG121L; EIG121-like; Estrogen-induced gene 121-like protein; h EIG121-like; hEIG121L; KIAA1324 like; KIAA1324L; KIAA1324-like; RIKEN cDNA 9330182L06 gene; UPF0577 protein KIAA1324-like; UPF0577 protein KIAA1324-like homolog
Gene Symbols KIAA1324L
Host Species Rabbit
Immunogen Recombinant Human KIAA1324L. Recombinant protein control fragment (Product #RP-108070).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222223
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.