Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA1328 Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144824
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144824 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144824 Supplier Invitrogen™ Supplier No. PA5144824
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AQALFKSREL VAEKQLTKPQ ELKLDMNGSD SGPSLLKSNC DGWLLGTSSS IKKHQDPPNS GENRKERKTV GFHSHMKDDA QW Antigen sequence identity: Mouse 60%; Rat 45%.

Competes with SMC1 for binding to SMC3. May affect the availability of SMC3 to engage in the formation of multimeric protein complexes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA1328
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KIAA1328
Gene Accession No. Q86T90
Gene Alias 4921538P22; AI461742; AW554918; expressed sequence AW554918; hinderin; KIAA1328; mKIAA1328; Protein hinderin; uncharacterized protein KIAA1328; uncharacterized protein KIAA1328; protein hinderin
Gene Symbols KIAA1328
Host Species Rabbit
Immunogen Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 130-211. Recombinant protein control fragment (Product #RP-109752).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57536
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Format Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.