Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA1429 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556772
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556772 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556772 Supplier Invitrogen™ Supplier No. PA556772
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSGRLDSDEQ KIQNDIIDIL LTFTQGVNEK LTISEETLAN NTWSLMLKEV LSSILKVPEG FFSGLILLSE LLPLPLPMQT TQVIEPHDIS VALNTRKL Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

KIAA1429 is required for N6-methyladenosine (m6A) methylation of mRNAs, a modification that plays a role in the efficiency of mRNA splicing and processing. It is also Involved in mRNA splicing regulation, probably via its function in m6A methylation. Diseases associated with KIAA1429 include Retinitis Pigmentosa 42.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA1429
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VIRMA
Gene Accession No. Q69YN4
Gene Alias 1110037F02Rik; 4930422M05Rik; DKFZP434I116; fSAP121; functional spliceosome-associated protein 121; KIAA1429; mKIAA1429; MSTP054; protein virilizer homolog; RGD1559904; RIKEN cDNA 1110037F02 gene; similar to mKIAA1429 protein; Virma
Gene Symbols VIRMA
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KIAA1429. Recombinant protein control fragment (Product #RP-93880).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25962
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.