Learn More
Invitrogen™ KIAA1429 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA595717
Description
Immunogen sequence: FMRQILNSDT IGCCGDDNGL MEVEGAHTSR TMSINAAELK QLLQSKEESP ENLFLELEKL VLEHSKDDDN LDSLLDSVVG LKQMLESSGD PLPLSDQDVE PVLSAPESLQ NLFNNRTAYV LADVMDDQLK SMWFTPFQAE EIDTDLDLVK VDLIELSEKC CSDFDLHSEL ERSFLSEPSS PGRTKTTKGF KLGKHKHETF ITSSGKSEYI EPAKRAHVVP PPRGRGRGGF GQGIRPHDIF RQRKQNTSRP PSMHVDDFVA AESKEVVPQD GIPPPKRPLK VSQKISSRGG FSGNRGGRGA F; Positive Samples: K-562, COS7, HeLa, PC-3.
KIAA1429 is required for N6-methyladenosine (m6A) methylation of mRNAs, a modification that plays a role in the efficiency of mRNA splicing and processing. It is also Involved in mRNA splicing regulation, probably via its function in m6A methylation. Diseases associated with KIAA1429 include Retinitis Pigmentosa 42.
Specifications
| KIAA1429 | |
| Polyclonal | |
| Unconjugated | |
| VIRMA | |
| 1110037F02Rik; 4930422M05Rik; DKFZP434I116; fSAP121; functional spliceosome-associated protein 121; KIAA1429; mKIAA1429; MSTP054; protein virilizer homolog; RGD1559904; RIKEN cDNA 1110037F02 gene; similar to mKIAA1429 protein; Virma | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 25962, 313061, 66185 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2.08 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| A2AIV2, Q69YN4 | |
| VIRMA | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1400-1700 of human KIAA1429 (NP_0563112). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.