Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA1429 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5144071
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5144071 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5144071 Supplier Invitrogen™ Supplier No. PA5144071
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human PC-3 whole cell, human SK-OV-3 whole cell, human K562 whole cell, human HEL whole cell. IHC: human bladder epithelial carcinoma tissue, human breast cancer tissue. ICC/IF: U20S cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

KIAA1429 is required for N6-methyladenosine (m6A) methylation of mRNAs, a modification that plays a role in the efficiency of mRNA splicing and processing. It is also Involved in mRNA splicing regulation, probably via its function in m6A methylation. Diseases associated with KIAA1429 include Retinitis Pigmentosa 42.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA1429
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene VIRMA
Gene Accession No. Q69YN4
Gene Alias 1110037F02Rik; 4930422M05Rik; DKFZP434I116; fSAP121; functional spliceosome-associated protein 121; KIAA1429; mKIAA1429; MSTP054; protein virilizer homolog; RGD1559904; RIKEN cDNA 1110037F02 gene; similar to mKIAA1429 protein; Virma
Gene Symbols VIRMA
Host Species Rabbit
Immunogen E.coli-derived human KIAA1429/VIRMA recombinant protein (Position: F11-K1529).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25962
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.