Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIAA1456 Polyclonal Antibody

KIAA1456 Polyclonal Antibody
Supplier: Thermo Scientific PA555150
Description
KIAA1456 Polyclonal Antibody for ICC/IF, IHC
Specifications
| KIAA1456 | |
| Polyclonal | |
| Unconjugated | |
| TRMT9B | |
| 6430573F11Rik; C27H8ORF79; C8orf79; KIAA1456; Probable tRNA methyltransferase 9B; probable tRNA methyltransferase 9-like protein; RIKEN cDNA 6430573F11 gene; RM9L; TRM9L; Trmt9b; Trmt9l; tRNA methyltransferase 9B (putative) | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 57604 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P272 | |
| TRMT9B | |
| Recombinant protein corresponding to Human KIAA1456. Recombinant protein control fragment (Product #RP-93899). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction