Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA1456 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109877
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109877 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109877 Supplier Invitrogen™ Supplier No. PA5109877
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VIHHFSTKQR RIRAIKEMAR VLVPGGQLMI YVWAMEQKNR HFEKQDVLVP WNRALCSQLF SESSQSGRKR QCGYPERGHP YHPPCSECSC SVCFKEQCGS KRSHSVGYEP AMARTCFANI SKEGEEEYGF YSTLGKSFRS WFFSRSLDES TLRKQIERVR PLKNTEVWAS STVTVQPSRH SSLDFDHQEP FSTKGQSLDE EVFVESSSGK HLEWLRAPGT L.

C8orf79 (chromosome 8 open reading frame 79) is a 454 amino acid protein that belongs to the methyltransferase superfamily and is encoded by a gene that maps to human chromosome 8p22. Consisting of nearly 146 million bases, chromosome 8 encodes about 800 genes. Translocation of portions of chromosome 8 with amplifications of the c-Myc gene are found in some leukemias and lymphomas, and are typically associated with a poor prognosis. Portions of chromosome 8 have been linked to schizophrenia and bipolar disorder.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA1456
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.24 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene TRMT9B
Gene Accession No. Q80WQ4, Q9P272
Gene Alias 6430573F11Rik; C27H8ORF79; C8orf79; KIAA1456; Probable tRNA methyltransferase 9B; probable tRNA methyltransferase 9-like protein; RIKEN cDNA 6430573F11 gene; RM9L; TRM9L; Trmt9b; Trmt9l; tRNA methyltransferase 9B (putative)
Gene Symbols TRMT9B
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 110-330 of human KIAA1456 (NP_065895.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 319582, 57604
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.