Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA1462 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545908
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545908 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545908 Supplier Invitrogen™ Supplier No. PA545908
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: QAARTGTRAG QGLQNGHEDG PAALAHRKTS AGKGHVSDSE SRRSTPRGHG Sequence homology: Human: 100%.

The function of this protein remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA1462
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene JCAD
Gene Accession No. Q9P266
Gene Alias JCAD; Junctional protein associated with coronary artery disease; KIAA1462
Gene Symbols JCAD
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human KIAA1462 (aa 31-80).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57608
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.