Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIAA1468 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558893
Description
Immunogen sequence: NVLLAKREEL IPLILCTACL HPEPKERDQL LHILFNLIKR PDDEQRQMIL TGCVAFARHV GPTRVEAELL PQCWEQINHK YPERRLLV Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Specifications
KIAA1468 | |
Polyclonal | |
Unconjugated | |
KIAA1468 | |
2310035C23Rik; 6430401N10; HsT3308; HsT885; Kiaa1468; lisH domain and HEAT repeat-containing protein KIAA1468; lisH domain and HEAT repeat-containing protein KIAA1468 homolog; mKIAA1468; RAB11 binding and LisH domain, coiled-coil and HEAT repeat-containing; RAB11-binding protein containing LisH, coiled-coil, and HEAT repeats; RAB11-binding protein RELCH; Relch; RGD1307235; RIKEN cDNA 2310035C23 gene; similar to RIKEN cDNA 2310035C23 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57614 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P260 | |
KIAA1468 | |
Recombinant protein corresponding to Human KIAA1468. Recombinant protein control fragment (Product #RP-98301). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction