Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIAA1468 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559039
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559039 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559039 Supplier Invitrogen™ Supplier No. PA559039
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EAYSALSCCF ISEDLMVNHF LPGLRCLRTD MEHLSPEHEV ILSSMIKECE QKVENKTVQE PQGSMSIAAS LVSEDTKTKF Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen KIAA1468
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KIAA1468
Gene Accession No. Q9P260
Gene Alias 2310035C23Rik; 6430401N10; HsT3308; HsT885; Kiaa1468; lisH domain and HEAT repeat-containing protein KIAA1468; lisH domain and HEAT repeat-containing protein KIAA1468 homolog; mKIAA1468; RAB11 binding and LisH domain, coiled-coil and HEAT repeat-containing; RAB11-binding protein containing LisH, coiled-coil, and HEAT repeats; RAB11-binding protein RELCH; Relch; RGD1307235; RIKEN cDNA 2310035C23 gene; similar to RIKEN cDNA 2310035C23
Gene Symbols KIAA1468
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KIAA1468. Recombinant protein control fragment (Product #RP-98302).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57614
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.