Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIAA1486 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA571132
Description
This target displays homology in the following species: Cow: 92%; Horse: 86%; Human: 100%; Rat: 79%.
Activates PI3K and concomitantly recruits the WAVE1 complex to the close vicinity of PI3K and regulates neuronal morphogenesis.
Specifications
KIAA1486 | |
Polyclonal | |
Unconjugated | |
NYAP2 | |
9430031J16Rik; Jr6; Kiaa1486; neuronal tyrosine phosphorylated adaptor for phosphoinositide 3-kinase adaptor 2; neuronal tyrosine-phophorylated phosphoinositide 3-kinase adaptor 2; neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adapter 2; neuronal tyrosine-phosphorylated phosphoinositide-3-kinase adaptor 2; NEWGENE_1305560; NYAP2 | |
Rabbit | |
Affinity chromatography | |
RUO | |
57624 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9P242 | |
NYAP2 | |
synthetic peptide directed towards the N-terminal region of human KIAA1486. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction