Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIAA1522 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566725
Description
Immunogen sequence: SAKAENDKHLS VGPGQGPGSA VDEHQDNVFF PSGRPPHLEE LHTQAQEGLR SLQHQEKQKL NKGGWDHGDT QSIQSSR Highest antigen sequence identity to the following orthologs - mouse 83%, rat 85%.
KIAA1522 gene ontology annotations related to this gene include cell differentiation.
Specifications
KIAA1522 | |
Polyclonal | |
Unconjugated | |
KIAA1522 | |
KIAA1522; uncharacterized protein KIAA1522 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
57648 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9P206 | |
KIAA1522 | |
Recombinant Human KIAA1522. Recombinant protein control fragment (Product #RP-107378). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction