Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIAA1524 Monoclonal Antibody (OTI2A1), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA808460

Description
Inhibition of protein phosphatase 2A (PP2A) activity has been identified as a prerequisite for the transformation of human cells. The protein, designated Cancerous Inhibitor of PP2A (CIP2A), interacts directly with the oncogenic transcription factor c-Myc, inhibits PP2A activity toward c-Myc serine 62 (S62), and thereby prevents c-Myc proteolytic degradation. In addition to its function in c-Myc stabilization, CIP2A promotes anchorage-independent cell growth and in vivo tumor formation. The oncogenic activity of CIP2A is demonstrated by transformation of human cells by overexpression of CIP2A. Importantly, CIP2A is overexpressed in two common human maligcies, head and neck squamous cell carcinoma (HNSCC) and colon cancer.Specifications
KIAA1524 | |
Monoclonal | |
Unconjugated | |
CIP2A | |
CIP2A | |
Human recombit protein fragment corresponding to amino acids 635-905 of human KIAA1524 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Western Blot | |
OTI2A1 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
KIAA1524, p90 | |
Mouse | |
Affinity Chromatography | |
RUO | |
57650 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction